This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04396875_ft
See other NCOA4-RET FUSION GE Assays ›
Gene Symbol:
NCOA4-RET fusion
Gene Name:
FUSION
Gene Aliases:
NCOA4, RET
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04396875_ft
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NCOA4-RET fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA OM_NCOA4-RET_N7R12.0 - 48 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Fusion Homo sapiens

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0