Assay Details
Gene Symbol: MIR223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029637.1 | 1 | 43 | 107 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | moesin | MSN | HEL70 | -276.9kb | Sense | Chr. X - 65588382 - 65741931 on NCBI Build GRCh38 |
Downstream | V-set and immunoglobulin domain containing 4 | VSIG4 |
CRIg, Z39IG |
2.8kb | Anti-sense | Chr. X - 66021738 - 66040125 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007478.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000356.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767803 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482453 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482440 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767765 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482459 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482466 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767798 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767769 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482457 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482443 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767770 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482439 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767753 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482477 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767764 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482465 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767750 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767753 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482465 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767796 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482466 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767764 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767798 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767765 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482443 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482457 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767776 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482459 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767769 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482477 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482440 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767795 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767770 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767803 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11767760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482453 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482439 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32482433 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 8167971 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004570.hg.1 | ENST00000385204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004570.hg.1 | ENST00000538676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004570.hg.1 | ENST00000540516.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004570.hg.1 | NR_029637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004570.hg.1 | TC0X000356.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000385204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007478.hg.1 | NR_029637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000385204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007478.hg.1 | NR_029637.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | ENST00000385204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | NR_029637.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | ENST00000385204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | ENST00000618234.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | ENST00000621933.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | NR_029637.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00151214.hg.1 | TC0X00007478.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3308936 | NR_029637.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3308936 | NR_029637.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308936 | NR_029637.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308936 | NR_029637.1 | human_transcriptome |
