This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04333119_s1
See other LINC01426 GE Assays ›
Gene Symbol:
LINC01426
Gene Name:
long intergenic non-protein coding RNA 1426
Gene Aliases:
lincRNA-uc002yug.2
Chromosome Location:
Chr.21: 34745825 - 34784886 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04333119_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01426
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038886.1 2 722 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 160 LINC00160 C21orf52,
NCRNA00160
-10.6kb Anti-sense Chr. 21 - 34723807 - 34737182 on NCBI Build GRCh38
Downstream runt related transcription factor 1 RUNX1 AML1,
AML1-EVI-1,
AMLCR1,
CBF2alpha,
CBFA2,
EVI-1,
PEBP2aB,
PEBP2alpha
39.4kb Anti-sense Chr. 21 - 34787801 - 35049334 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100007000.hg.1
This assay targets the same location as HTA transcript cluster(s) TC21000140.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR21001209.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR21001209.hg.1 NR_038886.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR21001209.hg.1 TC21000140.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 NR_038886.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 NR_038886.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007000.hg.1 NR_038886.1 Refseq
Affymetrix Human Clariom D Assay PSR2100147291.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100147291.hg.1 NR_038886.1 Refseq
Affymetrix Human Clariom D Assay PSR2100147291.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100147291.hg.1 NR_038886.1 Refseq
Affymetrix Human Clariom D Assay PSR2100147291.hg.1 TC2100007000.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2100147290.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100147290.hg.1 NR_038886.1 Refseq
Affymetrix Human Clariom D Assay PSR2100147290.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Clariom D Assay PSR2100147290.hg.1 NR_038886.1 Refseq
Affymetrix Human Clariom D Assay PSR2100147290.hg.1 TC2100007000.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000419921.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 ENST00000420877.1 Ensembl
Affymetrix Human Clariom D Assay TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Clariom D Assay TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Clariom D Assay TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Clariom D Assay TC2100007000.hg.1 NR_038885.1 Refseq
Affymetrix Human Clariom D Assay TC2100007000.hg.1 NR_038886.1 Refseq
Affymetrix Human Clariom D Assay TC2100007000.hg.1 NR_038886.1 Refseq
Affymetrix Human Clariom D Assay TC2100007000.hg.1 NR_038886.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0