This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332642_s1
See other MIR5192 GE Assays ›
Gene Symbol:
MIR5192
Gene Name:
microRNA 5192
Gene Aliases:
-
Chromosome Location:
Chr.2: 62205826 - 62205917 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332642_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR5192
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_049824.1 1 53 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream copper metabolism domain containing 1 COMMD1 C2orf5,
MURR1
-69.8kb Sense Chr. 2 - 61888723 - 62136070 on NCBI Build GRCh38
Downstream transmembrane protein 17 TMEM17 241.7kb Anti-sense Chr. 2 - 62452778 - 62535645 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0200007747.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200007747.hg.1 ENST00000579892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007747.hg.1 NR_049824.1 Refseq
Affymetrix Human Clariom D Assay PSR0200153488.hg.1 ENST00000579892.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153488.hg.1 NR_049824.1 Refseq
Affymetrix Human Clariom D Assay PSR0200153488.hg.1 TC0200007747.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200153488.hg.1 ENST00000579892.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200153488.hg.1 NR_049824.1 Refseq
Affymetrix Human Clariom D Assay TC0200007747.hg.1 ENST00000579892.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007747.hg.1 NR_049824.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0