Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007579.hg.1 |
ENST00000578054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007579.hg.1 |
NR_049809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152041.hg.1 |
ENST00000578054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152041.hg.1 |
NR_049809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152041.hg.1 |
ENST00000578054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152041.hg.1 |
NR_049809.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152041.hg.1 |
TC1800007579.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1800007579.hg.1 |
ENST00000578054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007579.hg.1 |
NR_049809.1 |
Refseq |