This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332520_s1
See other AIRN GE Assays ›
Gene Symbol:
AIRN
Gene Name:
antisense of IGF2R non-protein coding RNA
Gene Aliases:
AIR, IGF2R-AS, IGF2R-AS1, IGF2RAS, NCRNA00088
Chromosome Location:
Chr.6: 160003291 - 160007664 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332520_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIRN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_047514.1 1 3519 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MAS1 proto-oncogene,
G protein-coupled receptor
MAS1 MAS,
MGRA
-95.2kb Anti-sense Chr. 6 - 159906942 - 159908076 on NCBI Build GRCh38
Downstream calcineurin like EF-hand protein 1 pseudogene LOC729603 85.4kb Anti-sense Chr. 6 - 160093082 - 160096212 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600013767.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000609176.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000609176.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 ENST00000609176.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay PSR0600194944.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600194944.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay PSR0600194944.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600194944.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay PSR0600194944.hg.1 TC0600013767.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000609176.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000609176.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 ENST00000609176.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047511.1 Refseq
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047511.1 Refseq
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047511.1 Refseq
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay TC0600013767.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay PSR0600194942.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600194942.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay PSR0600194942.hg.1 ENST00000601203.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600194942.hg.1 NR_047514.1 Refseq
Affymetrix Human Clariom D Assay PSR0600194942.hg.1 TC0600013767.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header