This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04332414_m1
See other MAGI2-AS2 GE Assays ›
Gene Symbol:
MAGI2-AS2
Gene Name:
MAGI2 antisense RNA 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 79008988 - 79012277 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04332414_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAGI2-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046689.1 2 - 3 282 106
GenBank mRNA BX102296.1 - 2 - 3 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW117450.1 - 1 - 2 Chr. 7 - 79008990 - 79012277 [-] on Build GRCh38 106
GenBankmRNA AA971653.1 - 1 - 2 Chr. 7 - 79010927 - 79012277 [-] on Build GRCh38 106
GenBankmRNA AW665750.1 - 1 - 2 Chr. 7 - 79009094 - 79012277 [-] on Build GRCh38 106
GenBankmRNA AI218328.1 - 1 - 2 Chr. 7 - 79009030 - 79012277 [-] on Build GRCh38 106
GenBankmRNA AA884940.1 - 1 - 2 Chr. 7 - 79009043 - 79012277 [-] on Build GRCh38 106
GenBankmRNA AA972896.1 - 1 - 2 Chr. 7 - 79010950 - 79012277 [-] on Build GRCh38 106
GenBankmRNA AI684881.1 - 1 - 2 Chr. 7 - 79009024 - 79012277 [-] on Build GRCh38 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein L13a pseudogene 17 RPL13AP17 RPL13A_6_816 -651.5kb Sense Chr. 7 - 78347242 - 78359458 on NCBI Build GRCh38
Downstream MAGI2 antisense RNA 3 MAGI2-AS3 440.7kb Sense Chr. 7 - 79452957 - 79471208 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700008165.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700008165.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008165.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008165.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700008165.hg.1 NR_046689.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008165.hg.1 NR_046689.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700008165.hg.1 NR_046689.1 Refseq
Affymetrix Human Clariom D Assay TC0700008165.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008165.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008165.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Clariom D Assay TC0700008165.hg.1 NR_046689.1 Refseq
Affymetrix Human Clariom D Assay TC0700008165.hg.1 NR_046689.1 Refseq
Affymetrix Human Clariom D Assay TC0700008165.hg.1 NR_046689.1 Refseq
Affymetrix Human Clariom D Assay PSR0700156709.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700156709.hg.1 NR_046689.1 Refseq
Affymetrix Human Clariom D Assay PSR0700156709.hg.1 ENST00000411616.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700156709.hg.1 NR_046689.1 Refseq
Affymetrix Human Clariom D Assay PSR0700156709.hg.1 TC0700008165.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0