This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04331541_s1
See other MIR194-2 GE Assays ›
Gene Symbol:
MIR194-2 MIR194-2HG
Gene Name:
microRNA 194-2
MIR194-2 host gene
Gene Aliases:
MIRN194-2, mir-194-2
-
Chromosome Location:
Chr.11: 64891355 - 64891439 on Build GRCh38
Chromosome Location:
Chr.11: 64888458 - 64893449 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs04331541_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR194-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029829.1 1 43 70
Gene Symbol: MIR194-2HG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_133638.1 2 1810 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 192 MIR192 MIRN192,
miR-192,
miRNA192
-109.0 Sense Chr. 11 - 64891137 - 64891246 on NCBI Build GRCh38
Downstream autophagy related 2A ATG2A 3.1kb Sense Chr. 11 - 64894532 - 64917250 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1100013182.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001916.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841121 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841113 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841126 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841115 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841125 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841114 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841130 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841123 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841116 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841120 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841124 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841121 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841130 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841116 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841126 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841113 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841114 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841120 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841123 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841124 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841115 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07841125 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 7949275 - -
Affymetrix Human Transcriptome Array 2.0 TC1100013182.hg.1 ENST00000384864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013182.hg.1 ENST00000413053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013182.hg.1 ENST00000413053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013182.hg.1 NR_029829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11022909.hg.1 ENST00000384864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11022909.hg.1 NR_029829.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11022909.hg.1 TC11001916.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100204520.hg.1 ENST00000384864.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204520.hg.1 NR_029829.1 Refseq
Affymetrix Human Clariom D Assay PSR1100204520.hg.1 ENST00000384864.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204520.hg.1 NR_029829.1 Refseq
Affymetrix Human Clariom D Assay PSR1100204520.hg.1 TC1100013182.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100013182.hg.1 ENST00000384864.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013182.hg.1 ENST00000413053.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013182.hg.1 ENST00000413053.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013182.hg.1 NR_029829.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309955 NR_029829.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309955 NR_029829.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309955 NR_029829.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309955 NR_029829.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0