This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04277464_s1
See other LOC100505549 GE Assays ›
Gene Symbol:
LOC100505549
Gene Name:
uncharacterized LOC100505549
Gene Aliases:
-
Chromosome Location:
Chr.18: 57630302 - 57669276 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04277464_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100505549
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001242804.1 NP_001229733.1 3 1119 125
GenBank mRNA AK058035.1 - 1 1881 125
BC040476.1 - 3 1072 125

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800009232.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000196.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human CGH Microarray 4x180K 149584 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 868308 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 374696 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 385593 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 103727 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 20481 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 43056 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 348878 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 142418 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 399779 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 333279 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 306876 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 376814 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 326565 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 321899 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 374690 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 209521 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 149724 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 358717 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 355808 - -
Agilent Human Genome CGH + SNP Microarray A_16_P03377213 - -
Agilent Human Genome CGH + SNP Microarray A_16_P41055671 - -
Agilent Human Genome CGH + SNP Microarray A_16_P41055671 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P41055671 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P41055671 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03377229 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 126806 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 57476 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12900023 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41055663 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20877094 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41055671 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20877087 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P41055671 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P20877094 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 8021366 - -
Affymetrix Human Transcriptome Array 2.0 PSR18002307.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002307.hg.1 TC18000196.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR18002310.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002310.hg.1 TC18000196.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002309.hg.1 NM_001242804.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18002309.hg.1 TC18000196.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1800163621.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163621.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163621.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163621.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163621.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163621.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163621.hg.1 TC1800009232.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Clariom D Assay TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay TC1800009232.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163625.hg.1 TC1800009232.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163623.hg.1 TC1800009232.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800163628.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163628.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163628.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163628.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163628.hg.1 ENST00000619899.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163628.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163628.hg.1 TC1800009232.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800163629.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163629.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163629.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163629.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163629.hg.1 TC1800009232.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 ENST00000591854.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 ENST00000592201.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 NM_001242804.1 Refseq
Affymetrix Human Clariom D Assay PSR1800163624.hg.1 TC1800009232.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 109362 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0