Assay Details
Gene Symbol: IQCH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040055.1 | 1 | 119 | 112 | ||
GenBank mRNA | AW204237.1 | - | 3 | 112 | ||
BX093700.1 | - | 1 | 115 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | IQ motif containing H | IQCH | NYDSP5 | -14.2kb | Anti-sense | Chr. 15 - 67254772 - 67501804 on NCBI Build GRCh38 |
Downstream | chromosome 15 open reading frame 61 | C15orf61 | 211.0 | Anti-sense | Chr. 15 - 67521184 - 67527303 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001588.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15015299.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15015299.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15015299.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15015299.hg.1 | TC15001588.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15015304.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15015304.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15015304.hg.1 | TC15001588.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173066.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500173066.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173066.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500173066.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173066.hg.1 | TC1500009848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173060.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173060.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173060.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173060.hg.1 | NR_040054.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173060.hg.1 | NR_040055.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173060.hg.1 | NR_040056.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500173060.hg.1 | TC1500009848.hg.1 | Clariom D TC |
