Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009418.hg.1 |
NR_039882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009418.hg.1 |
TC17000723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008457.hg.1 |
ENST00000584140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008457.hg.1 |
NR_039882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008457.hg.1 |
ENST00000584140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008457.hg.1 |
NR_039882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162953.hg.1 |
ENST00000584140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162953.hg.1 |
NR_039882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162953.hg.1 |
ENST00000584140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162953.hg.1 |
NR_039882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162953.hg.1 |
TC1700008457.hg.1 |
Clariom D TC |