This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04274600_s1
See other MIR3975 GE Assays ›
Gene Symbol:
MIR3975
Gene Name:
microRNA 3975
Gene Aliases:
-
Chromosome Location:
Chr.18: 35591737 - 35591806 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04274600_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR3975
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_039771.1 1 34 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream INO80 complex subunit C INO80C C18orf37,
IES6,
hIes6
-93.7kb Anti-sense Chr. 18 - 35468327 - 35497991 on NCBI Build GRCh38
Downstream polypeptide N-acetylgalactosaminyltransferase 1 GALNT1 GALNAC-T1 35.6kb Sense Chr. 18 - 35581066 - 35711834 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1800007081.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000146.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR18001660.hg.1 NR_039771.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18001660.hg.1 TC18000146.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1800148574.hg.1 NR_039771.1 Refseq
Affymetrix Human Clariom D Assay PSR1800148574.hg.1 NR_039771.1 Refseq
Affymetrix Human Clariom D Assay PSR1800148574.hg.1 TC1800007081.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header