Assay Details
Gene Symbol: LOC100128993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038919.1 | 3 - 4 | 565 | 59 | ||
GenBank mRNA | DA928996.1 | - | 2 - 3 | 277 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pleckstrin and Sec7 domain containing 3 | PSD3 |
EFA6D, EFA6R, HCA67 |
-97.7kb | Sense | Chr. 8 - 18527303 - 19086909 on NCBI Build GRCh38 |
Downstream | SH2 domain containing 4A | SH2D4A |
PPP1R38, SH2A |
86.2kb | Anti-sense | Chr. 8 - 19313443 - 19396218 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001019.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08012949.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012949.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012949.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012949.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08012949.hg.1 | TC08001019.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08012946.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012946.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08012946.hg.1 | TC08001019.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08012951.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012951.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08012951.hg.1 | TC08001019.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08012952.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08012952.hg.1 | TC08001019.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08012947.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012947.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08012947.hg.1 | TC08001019.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08012948.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012948.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012948.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012948.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08012948.hg.1 | TC08001019.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0800166435.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166435.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166435.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166435.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166435.hg.1 | TC0800009774.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166439.hg.1 | TC0800009774.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800166444.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166444.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166444.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166444.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166444.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166444.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166444.hg.1 | TC0800009774.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800166433.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166433.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166433.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166433.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166433.hg.1 | TC0800009774.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166438.hg.1 | TC0800009774.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000517949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | ENST00000520920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009774.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166434.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166434.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166434.hg.1 | ENST00000518417.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166434.hg.1 | NR_038919.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166434.hg.1 | TC0800009774.hg.1 | Clariom D TC |
