Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007019.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002428.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002428.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002428.hg.1 |
TC08000186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002425.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002425.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002425.hg.1 |
TC08000186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002426.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002426.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08002426.hg.1 |
TC08000186.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0800148052.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148052.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148052.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148052.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148052.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148052.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148052.hg.1 |
TC0800007019.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007019.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148055.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148055.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148055.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148055.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148055.hg.1 |
TC0800007019.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800148053.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148053.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148053.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148053.hg.1 |
ENST00000517420.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148053.hg.1 |
ENST00000519692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800148053.hg.1 |
NR_038323.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800148053.hg.1 |
TC0800007019.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
1562263_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1562263_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1562263_at6 |
- |
- |