This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273432_s1
See other MIR1207 GE Assays ›
Gene Symbol:
MIR1207
Gene Name:
microRNA 1207
Gene Aliases:
MIRN1207, hsa-mir-1207
Chromosome Location:
Chr.8: 128049152 - 128049238 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273432_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR1207
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031612.1 1 44 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1206 MIR1206 MIRN1206,
hsa-mir-1206
-40.2kb Sense Chr. 8 - 128008898 - 128008956 on NCBI Build GRCh38
Downstream microRNA 1208 MIR1208 MIRN1208,
hsa-mir-1208
100.9kb Sense Chr. 8 - 128150116 - 128150188 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0800008858.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000755.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744806 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744797 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744818 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462681 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462687 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744823 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462683 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462718 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462719 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462715 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744790 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744795 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462707 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744801 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744788 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462695 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462701 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462697 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744783 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744814 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744802 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462690 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16134549 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462696 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744791 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462712 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462713 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462709 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462698 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462693 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744821 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744787 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462687 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744806 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462681 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744790 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462695 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16134549 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744823 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462713 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462701 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462697 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462709 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744797 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462690 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744788 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462715 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462698 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744795 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744818 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744783 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744821 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462707 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462696 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744801 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744814 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744802 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462718 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744787 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26744791 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462683 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462693 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462719 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06462712 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800008858.hg.1 ENST00000408249.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800008858.hg.1 NR_031612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08010118.hg.1 ENST00000408249.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08010118.hg.1 NR_031612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08010118.hg.1 TC08000755.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0800008858.hg.1 ENST00000408249.1 Ensembl
Affymetrix Human Clariom D Assay TC0800008858.hg.1 NR_031612.1 Refseq
Affymetrix Human Clariom D Assay PSR0800160649.hg.1 ENST00000408249.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800160649.hg.1 NR_031612.1 Refseq
Affymetrix Human Clariom D Assay PSR0800160649.hg.1 ENST00000408249.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800160649.hg.1 NR_031612.1 Refseq
Affymetrix Human Clariom D Assay PSR0800160649.hg.1 TC0800008858.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308758 NR_031612.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308758 NR_031612.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308758 NR_031612.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308758 NR_031612.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm