This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273424_s1
See other MIR1231 GE Assays ›
Gene Symbol:
MIR1231
Gene Name:
microRNA 1231
Gene Aliases:
MIRN1231, hsa-mir-1231
Chromosome Location:
Chr.1: 201808611 - 201808702 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273424_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR1231
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031599.1 1 51 57

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream IPO9 antisense RNA 1 IPO9-AS1 Anti-sense Chr. 1 - 201688256 - 201829559 on NCBI Build GRCh38
Downstream importin 9 IPO9 Imp9 20.5kb Sense Chr. 1 - 201829160 - 201884294 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0100011191.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001663.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790570 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790589 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790566 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730419 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790590 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15162097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730416 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790602 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730401 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730407 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15162094 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790584 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790575 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730418 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790591 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730423 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730409 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790598 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790573 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790585 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730387 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730397 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730425 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730402 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790571 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730408 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730389 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790580 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730399 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790572 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790601 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730427 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730404 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730416 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790584 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15162094 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790573 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730399 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730387 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730427 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730407 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790589 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790570 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730389 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15162097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790566 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790590 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730401 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790591 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730425 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730423 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790601 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730408 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790572 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730404 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730418 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790575 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790580 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730397 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730419 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790571 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730409 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790602 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790598 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00730402 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20790585 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100011191.hg.1 ENST00000408101.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011191.hg.1 NR_031599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01025744.hg.1 ENST00000408101.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01025744.hg.1 NR_031599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01025744.hg.1 TC01001663.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100182029.hg.1 ENST00000408101.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100182029.hg.1 NR_031599.1 Refseq
Affymetrix Human Clariom D Assay PSR0100182029.hg.1 ENST00000408101.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100182029.hg.1 NR_031599.1 Refseq
Affymetrix Human Clariom D Assay PSR0100182029.hg.1 TC0100011191.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100011191.hg.1 ENST00000408101.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011191.hg.1 NR_031599.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308520 NR_031599.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308520 NR_031599.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0