This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04273257_s1
See other MIR101-2 GE Assays ›
Gene Symbol:
MIR101-2
Gene Name:
microRNA 101-2
Gene Aliases:
MIRN101-2, mir-101-2
Chromosome Location:
Chr.9: 4850297 - 4850375 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04273257_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR101-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029836.1 1 35 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream adenylate kinase 3 AK3 AK3L1,
AK6,
AKL3L,
AKL3L1,
FIX
-108.3kb Anti-sense Chr. 9 - 4709556 - 4742043 on NCBI Build GRCh38
Downstream Janus kinase 2 JAK2 JTK10,
THCYT3
134.9kb Sense Chr. 9 - 4985086 - 5128183 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0900006532.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000033.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560779 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560765 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848939 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848909 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560785 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848875 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848915 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848891 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560784 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560807 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560782 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848885 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848912 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848921 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848884 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560778 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848873 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848919 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848876 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848904 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848861 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848888 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848928 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560759 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848890 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560750 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560780 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848927 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560813 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560826 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848867 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560745 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848906 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560798 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560776 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848913 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848880 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848905 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848941 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560820 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848893 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848886 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848914 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848911 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848903 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560747 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560792 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848929 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848935 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848887 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848933 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848918 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848900 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560802 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560763 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848877 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560790 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560807 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560779 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848877 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848880 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560798 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848884 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848913 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848861 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848905 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848912 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560813 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848903 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560765 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848928 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848887 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560778 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848927 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848921 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848914 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848885 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560792 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560790 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848919 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560759 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560802 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560784 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560780 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560826 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848941 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848918 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560742 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560785 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848906 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848911 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560750 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848929 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848904 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848876 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560745 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560782 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848909 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848875 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848873 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560820 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848915 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848891 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560763 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560776 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848890 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06560747 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848867 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848939 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848935 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848886 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848933 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848900 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848888 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26848893 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 8154177 - -
Affymetrix Human Gene 1.1 ST Array 8154176 - -
Affymetrix Human Transcriptome Array 2.0 TC0900006532.hg.1 ENST00000362195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006532.hg.1 NR_029836.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09000421.hg.1 ENST00000362195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09000421.hg.1 NR_029836.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09000421.hg.1 TC09000033.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0900009443.hg.1 ENST00000443970.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009443.hg.1 ENST00000443970.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006532.hg.1 ENST00000362195.2 Ensembl
Affymetrix Human Clariom D Assay TC0900006532.hg.1 NR_029836.1 Refseq
Affymetrix Human Clariom D Assay PSR0900144794.hg.1 ENST00000362195.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900144794.hg.1 NR_029836.1 Refseq
Affymetrix Human Clariom D Assay PSR0900144794.hg.1 ENST00000362195.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900144794.hg.1 NR_029836.1 Refseq
Affymetrix Human Clariom D Assay PSR0900144794.hg.1 TC0900006532.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308946 NR_029836.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3308946 AF480540.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308946 NR_029836.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308946 AF480540.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308946 NR_029836.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308946 AF480540.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308946 NR_029836.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308946 AF480540.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0