This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04260261_s1
See other GPR183 GE Assays ›
Gene Symbol:
GPR183
Gene Name:
G protein-coupled receptor 183
Gene Aliases:
EBI2
Chromosome Location:
Chr.13: 99294273 - 99307495 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04260261_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR183
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004951.4 NP_004942.1 2 1220 83
XM_017020405.1 XP_016875894.1 2 1163 83
GenBank mRNA AI740732.1 - 1 83
AK223390.1 - 2 1124 83
AK292091.1 - 2 1144 83
AW574697.1 - 1 83
BC020752.1 - 2 1124 4722289 83
L08177.1 - 1 1079 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009598.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000829.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR183

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
B cell activation involved in immune response
mature B cell differentiation involved in immune response
dendritic cell chemotaxis
immune response
humoral immune response
G-protein coupled receptor signaling pathway
T cell chemotaxis
osteoclast differentiation
leukocyte chemotaxis
positive regulation of B cell proliferation
dendritic cell homeostasis
T follicular helper cell differentiation
positive regulation of ERK1 and ERK2 cascade
regulation of astrocyte chemotaxis
G-protein coupled receptor activity
oxysterol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13009345.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009345.hg.1 NM_004951.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009345.hg.1 TC13000829.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300164221.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164221.hg.1 NM_004951.4 Refseq
Affymetrix Human Clariom D Assay PSR1300164221.hg.1 ENST00000376414.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164221.hg.1 NM_004951.4 Refseq
Affymetrix Human Clariom D Assay PSR1300164221.hg.1 TC1300009598.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0