This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04259679_s1
See other CITED4 GE Assays ›
Gene Symbol:
CITED4
Gene Name:
Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 40861056 - 40862346 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04259679_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CITED4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133467.2 NP_597724.1 1 653 100
GenBank mRNA BC035496.2 - 1 655 3622207 100
BC052559.1 - 1 653 6250908 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100013846.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CITED4

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
developmental process
response to estrogen
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription coactivator activity
transcription coactivator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100013846.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013846.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013846.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013846.hg.1 NM_133467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013846.hg.1 NM_133467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013846.hg.1 NM_133467.2 Refseq
Affymetrix Human Clariom D Assay TC0100013846.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013846.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013846.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Clariom D Assay TC0100013846.hg.1 NM_133467.2 Refseq
Affymetrix Human Clariom D Assay TC0100013846.hg.1 NM_133467.2 Refseq
Affymetrix Human Clariom D Assay TC0100013846.hg.1 NM_133467.2 Refseq
Affymetrix Human Clariom D Assay PSR0100203541.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100203541.hg.1 NM_133467.2 Refseq
Affymetrix Human Clariom D Assay PSR0100203541.hg.1 ENST00000372638.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100203541.hg.1 NM_133467.2 Refseq
Affymetrix Human Clariom D Assay PSR0100203541.hg.1 TC0100013846.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0