This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04233529_s1
See other MIR320A GE Assays ›
Gene Symbol:
MIR320A
Gene Name:
microRNA 320a
Gene Aliases:
MIRN320, MIRN320A, hsa-mir-320a, mir-320a
Chromosome Location:
Chr.8: 22244962 - 22245043 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04233529_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR320A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029714.1 1 42 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phytanoyl-CoA 2-hydroxylase interacting protein PHYHIP DYRK1AP3,
PAHX-AP,
PAHXAP1
-12.6kb Sense Chr. 8 - 22219703 - 22232338 on NCBI Build GRCh38
Downstream RNA polymerase III subunit D POLR3D BN51T,
RPC4,
RPC53,
TSBN51
61.0 Anti-sense Chr. 8 - 22245104 - 22251167 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0800009834.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001037.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287024 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287027 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287029 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287026 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06005145 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287028 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06005146 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06005138 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287027 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287029 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287024 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06005138 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287028 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26287026 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06005145 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06005146 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 8149705 - -
Affymetrix Human Transcriptome Array 2.0 TC0800009834.hg.1 ENST00000385302.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009834.hg.1 NR_029714.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08013247.hg.1 ENST00000385302.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08013247.hg.1 NR_029714.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08013247.hg.1 TC08001037.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0800166821.hg.1 ENST00000385302.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800166821.hg.1 NR_029714.1 Refseq
Affymetrix Human Clariom D Assay PSR0800166821.hg.1 ENST00000385302.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800166821.hg.1 NR_029714.1 Refseq
Affymetrix Human Clariom D Assay PSR0800166821.hg.1 TC0800009834.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800009834.hg.1 ENST00000385302.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009834.hg.1 NR_029714.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309099 NR_029714.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309099 NR_029714.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309099 NR_029714.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309099 NR_029714.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0