This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04233526_s1
See other MIR214 GE Assays ›
Gene Symbol:
MIR214 DNM3OS
Gene Name:
microRNA 214
DNM3 opposite strand/antisense RNA
Gene Aliases:
MIRN214, miRNA214, mir-214
DNM3-AS1, MIR199A2HG
Chromosome Location:
Chr.1: 172138798 - 172138907 on Build GRCh38
Chromosome Location:
Chr.1: 172136879 - 172144835 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs04233526_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR214
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029627.1 1 53 85
Gene Symbol: DNM3OS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038397.2 1 5981 85
NR_103486.1 4 2075 85
GenBank mRNA FJ623959.1 - 1 5981 85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CF529290.1 - 1 Chr. 1 - 172138581 - 172139062 [+] on Build GRCh38 85
GenBankmRNA BQ012655.1 - 1 Chr. 1 - 172138581 - 172138936 [+] on Build GRCh38 85
GenBankmRNA BQ447950.1 - 1 Chr. 1 - 172138585 - 172139284 [+] on Build GRCh38 85
GenBankmRNA CA439854.1 - 1 286 Chr. 1 - 172138578 - 172139131 [-] on Build GRCh38 85
GenBankmRNA AI039268.1 - 1 Chr. 1 - 172138581 - 172139002 [+] on Build GRCh38 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DNM3 intronic transcript 1 DNM3-IT1 -274.1kb Anti-sense Chr. 1 - 171864192 - 171864687 on NCBI Build GRCh38
Downstream microRNA 199a-2 MIR199A2 MIR-199-s,
MIRN199A2,
mir-199a-2
5.6kb Sense Chr. 1 - 172144535 - 172144644 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0100016414.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003519.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Oligo Microarray 4x44K 8340 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597494 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597461 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656925 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597473 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656897 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656914 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656913 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656892 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656899 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597500 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656905 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656927 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656928 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656931 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656895 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656889 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656918 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656922 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597471 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656908 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656932 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656923 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597444 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656907 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656890 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656890 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656927 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656913 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597473 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656931 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597471 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656907 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656925 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597444 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656892 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656905 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656922 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597494 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656899 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656914 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597461 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656932 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656889 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656895 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656897 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656908 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656918 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656923 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00597500 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20656928 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 7922329 - -
Affymetrix Human Gene 1.0 ST Array 7922327 - -
Affymetrix Human Gene 1.1 ST Array 7922326 - -
Affymetrix HT HG-U133+ PM Array Plate 235891_PM_at - -
Affymetrix Human Transcriptome Array 2.0 PSR01054619.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01054619.hg.1 TC01003519.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01054620.hg.1 ENST00000385214.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01054620.hg.1 NR_029627.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01054620.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01054620.hg.1 TC01003519.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000385214.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000385289.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_029618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_029627.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 ENST00000385214.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 NR_029627.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 ENST00000385214.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 NR_029627.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224744.hg.1 TC0100016414.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000385214.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000385289.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 ENST00000417354.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_029618.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_029627.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay TC0100016414.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224740.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224740.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224740.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224740.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224740.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224740.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224740.hg.1 TC0100016414.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100224745.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224745.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224745.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224745.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224745.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224745.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224745.hg.1 TC0100016414.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100224743.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224743.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224743.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224743.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224743.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224743.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224743.hg.1 TC0100016414.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100224741.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224741.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224741.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224741.hg.1 ENST00000417354.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100224741.hg.1 NR_038397.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224741.hg.1 NR_103486.1 Refseq
Affymetrix Human Clariom D Assay PSR0100224741.hg.1 TC0100016414.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A007925 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A007925 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 205880 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308597 NR_038397.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3308597 FJ623959.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3308597 NR_029627.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308597 NR_038397.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308597 FJ623959.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308597 NR_029627.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308597 NR_038397.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308597 FJ623959.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308597 NR_029627.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308597 NR_038397.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308597 FJ623959.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308597 NR_029627.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0