This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04232243_s1
See other GAS5-AS1 GE Assays ›
Gene Symbol:
GAS5-AS1
Gene Name:
GAS5 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 173863248 - 173863941 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04232243_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GAS5-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037605.1 1 145 75
GenBank mRNA AF088026.1 - 1 145 75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA W39642.1 - 144 Chr. - - [-] on Build 75
GenBankmRNA DA168988.1 - 1 47 Chr. 1 - 173863346 - 173863908 [+] on Build GRCh38 75
GenBankmRNA BQ184822.1 - 1 Chr. 1 - 173863253 - 173863941 [-] on Build GRCh38 75
GenBankmRNA AA296410.1 - 1 135 Chr. 1 - 173863258 - 173863578 [+] on Build GRCh38 75
GenBankmRNA BX102112.1 - 1 145 Chr. 1 - 173863248 - 173863941 [+] on Build GRCh38 75
GenBankmRNA BQ187737.1 - 2 304 Chr. 1 - 173862152 - 173863731 [+] on Build GRCh38 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream growth arrest specific 5 (non-protein coding) GAS5 NCRNA00030,
SNHG2
Anti-sense Chr. 1 - 173863901 - 173867987 on NCBI Build GRCh38
Downstream small nucleolar RNA,
C/D box 81
SNORD81 RNU104,
U81,
Z23
234.0 Anti-sense Chr. 1 - 173864175 - 173864217 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100010662.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001517.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar

Array Name Probe IDs Targets Target Type
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P307211 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 29974 - -
Whole Human Genome Oligo Microarray G4112A A_32_P307211 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P307211 - -
Agilent Human 1B Microarray G4111A A_32_P307211 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100010662.hg.1 ENST00000602767.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010662.hg.1 NR_037605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01023457.hg.1 ENST00000416578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01023457.hg.1 NR_037605.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01023457.hg.1 TC01001517.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100178388.hg.1 ENST00000602767.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100178388.hg.1 NR_037605.1 Refseq
Affymetrix Human Clariom D Assay PSR0100178388.hg.1 ENST00000602767.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100178388.hg.1 NR_037605.1 Refseq
Affymetrix Human Clariom D Assay PSR0100178388.hg.1 TC0100010662.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100010662.hg.1 ENST00000602767.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010662.hg.1 NR_037605.1 Refseq
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A035273 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A035273 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header