Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007339.hg.1 |
ENST00000585069.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007339.hg.1 |
NR_037508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004033.hg.1 |
NR_037508.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004033.hg.1 |
TC07000263.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0700007339.hg.1 |
ENST00000585069.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007339.hg.1 |
NR_037508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150895.hg.1 |
ENST00000585069.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150895.hg.1 |
NR_037508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150895.hg.1 |
ENST00000585069.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150895.hg.1 |
NR_037508.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150895.hg.1 |
TC0700007339.hg.1 |
Clariom D TC |