This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04232171_s1
See other MIR23C GE Assays ›
Gene Symbol:
MIR23C
Gene Name:
microRNA 23c
Gene Aliases:
hsa-mir-23c, mir-23c
Chromosome Location:
Chr.X: 20017088 - 20017187 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04232171_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR23C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037414.1 1 53 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MAP7 domain containing 2 MAP7D2 Sense Chr. X - 20006713 - 20116996 on NCBI Build GRCh38
Downstream eukaryotic translation initiation factor 1A,
X-linked
EIF1AX EIF1A,
EIF1AP1,
EIF4C,
eIF-1A,
eIF-4C
107.3kb Sense Chr. X - 20124518 - 20141848 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0X00009214.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000908.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00009214.hg.1 ENST00000579846.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009214.hg.1 NR_037414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X012279.hg.1 NR_037414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X012279.hg.1 TC0X000908.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0X00163331.hg.1 ENST00000579846.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00163331.hg.1 NR_037414.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00163331.hg.1 ENST00000579846.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00163331.hg.1 NR_037414.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00163331.hg.1 TC0X00009214.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00009214.hg.1 ENST00000579846.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009214.hg.1 NR_037414.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0