This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04232167_s1
See other MIR3616 GE Assays ›
Gene Symbol:
MIR3616
Gene Name:
microRNA 3616
Gene Aliases:
mir-3616
Chromosome Location:
Chr.20: 47166967 - 47167058 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04232167_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR3616
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037410.1 1 48 66

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 2 member 10 SLC2A10 ATS,
GLUT10
-430.6kb Sense Chr. 20 - 46708358 - 46736347 on NCBI Build GRCh38
Downstream zinc finger MYND-type containing 8 ZMYND8 PRKCBP1,
PRO2893,
RACK7
42.2kb Anti-sense Chr. 20 - 47209214 - 47357780 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC2000007552.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000369.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000007552.hg.1 ENST00000584070.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007552.hg.1 NR_037410.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005386.hg.1 NR_037410.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005386.hg.1 TC20000369.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC2000007552.hg.1 ENST00000584070.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007552.hg.1 NR_037410.1 Refseq
Affymetrix Human Clariom D Assay PSR2000152726.hg.1 ENST00000584070.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000152726.hg.1 NR_037410.1 Refseq
Affymetrix Human Clariom D Assay PSR2000152726.hg.1 ENST00000584070.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000152726.hg.1 NR_037410.1 Refseq
Affymetrix Human Clariom D Assay PSR2000152726.hg.1 TC2000007552.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0