Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004988.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004988.hg.1 |
TC08000342.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
ENST00000521874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004989.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004989.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004989.hg.1 |
TC08000342.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0800151998.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151998.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151998.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151998.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151998.hg.1 |
TC0800007527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800151993.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151993.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151993.hg.1 |
TC0800007527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800151992.hg.1 |
ENST00000521874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151992.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151992.hg.1 |
ENST00000521874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151992.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151992.hg.1 |
TC0800007527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800151997.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151997.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151997.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151997.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151997.hg.1 |
TC0800007527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800151995.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151995.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151995.hg.1 |
TC0800007527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800151996.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151996.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151996.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151996.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151996.hg.1 |
TC0800007527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
ENST00000521874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
ENST00000528139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007527.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151994.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151994.hg.1 |
NR_037168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151994.hg.1 |
TC0800007527.hg.1 |
Clariom D TC |
Human Genome U95 Set |
63707_r_at |
- |
- |
Human Genome U95 Set |
63707_PM_r_at |
- |
- |