This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231809_s1
See other MIR3166 GE Assays ›
Gene Symbol:
MIR3166
Gene Name:
microRNA 3166
Gene Aliases:
-
Chromosome Location:
Chr.11: 88176502 - 88176593 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231809_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR3166
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036124.1 1 48 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RAB38,
member RAS oncogene family
RAB38 NY-MEL-1,
rrGTPbp
-1.0kb Anti-sense Chr. 11 - 87809490 - 88175505 on NCBI Build GRCh38
Downstream cathepsin C CTSC CPPI,
DPP-I,
DPP1,
DPPI,
HMS,
JP,
JPD,
PALS,
PDON1,
PLS
117.0kb Anti-sense Chr. 11 - 88293592 - 88337787 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1100008696.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100008696.hg.1 ENST00000577344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008696.hg.1 NR_036124.1 Refseq
Affymetrix Human Clariom D Assay TC1100008696.hg.1 ENST00000577344.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008696.hg.1 NR_036124.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164291.hg.1 ENST00000577344.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164291.hg.1 NR_036124.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164291.hg.1 ENST00000577344.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100164291.hg.1 NR_036124.1 Refseq
Affymetrix Human Clariom D Assay PSR1100164291.hg.1 TC1100008696.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0