This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231689_s1
See other MIR590 GE Assays ›
Gene Symbol:
MIR590
Gene Name:
microRNA 590
Gene Aliases:
MIRN590, hsa-mir-590, mir-590
Chromosome Location:
Chr.7: 74191198 - 74191294 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231689_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR590
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030321.1 1 40 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW812109.1 - 1 Chr. 7 - 74191002 - 74191344 [-] on Build GRCh38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream eukaryotic translation initiation factor 4H EIF4H WBSCR1,
WSCR1,
eIF-4H
Sense Chr. 7 - 74174376 - 74197101 on NCBI Build GRCh38
Downstream linker for activation of T-cells family member 2 LAT2 HSPC046,
LAB,
NTAL,
WBSCR15,
WBSCR5,
WSCR5
18.1kb Sense Chr. 7 - 74209757 - 74229834 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0700007999.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000452.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535468 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535438 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779314 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535404 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779284 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535448 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779285 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535429 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535460 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779299 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535405 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779327 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779277 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535434 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535435 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535420 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535451 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779312 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779335 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535423 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779303 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779305 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779332 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535430 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779296 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779301 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535414 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535407 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779324 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535462 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535445 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535454 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779293 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779322 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535457 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779290 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535409 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779316 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535435 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779303 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779324 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535468 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535462 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779284 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779277 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779301 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779332 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535457 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535430 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779293 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779290 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535445 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779314 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535420 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779322 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779335 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535429 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779316 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779299 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535434 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535423 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535409 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779327 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535404 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535448 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779313 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535407 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535405 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535454 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779296 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535414 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779285 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535451 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535438 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779312 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25779305 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05535460 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07006118.hg.1 ENST00000385008.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07006118.hg.1 NR_030321.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07006118.hg.1 TC07000452.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0700007999.hg.1 ENST00000385008.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007999.hg.1 NR_030321.1 Refseq
Affymetrix Human Clariom D Assay TC0700007999.hg.1 ENST00000385008.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007999.hg.1 NR_030321.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155251.hg.1 ENST00000385008.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700155251.hg.1 NR_030321.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155251.hg.1 ENST00000385008.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700155251.hg.1 NR_030321.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155251.hg.1 TC0700007999.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308345 NR_030321.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308345 NR_030321.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308345 NR_030321.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308345 NR_030321.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0