This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231684_s1
See other MIR585 GE Assays ›
Gene Symbol:
MIR585
Gene Name:
microRNA 585
Gene Aliases:
MIRN585, hsa-mir-585, mir-585
Chromosome Location:
Chr.5: 169263601 - 169263694 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231684_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR585
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030311.1 1 43 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC728095 LOC728095 -225.6kb Anti-sense Chr. 5 - 169013227 - 169037998 on NCBI Build GRCh38
Downstream spindle apparatus coiled-coil protein 1 SPDL1 CCDC99 319.9kb Anti-sense Chr. 5 - 169583634 - 169604778 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0500012766.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05002046.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633764 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633767 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454832 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454836 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454856 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633765 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454877 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633769 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454838 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454873 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633799 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633766 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633761 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454866 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633809 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454833 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633807 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454847 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454841 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633773 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633789 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454843 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454850 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454871 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633780 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633818 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633762 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633798 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454861 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633791 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633801 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454844 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633770 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454823 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454839 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633792 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454870 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454853 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633815 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454874 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454848 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454865 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454849 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454849 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454847 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633789 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633780 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454877 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454841 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633766 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633792 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454838 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454823 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633762 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454865 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633769 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633773 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454844 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454866 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633818 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633770 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454873 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633809 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454874 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454848 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454850 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633791 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454832 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454839 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454843 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454871 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633765 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454833 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633815 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633764 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633761 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633801 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633798 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633799 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454853 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633767 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454870 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454836 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24633807 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454861 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04454856 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05028199.hg.1 ENST00000384887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05028199.hg.1 NR_030311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05028199.hg.1 TC05002046.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500012766.hg.1 ENST00000384887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012766.hg.1 NR_030311.1 Refseq
Affymetrix Human Clariom D Assay TC0500012766.hg.1 ENST00000384887.1 Ensembl
Affymetrix Human Clariom D Assay TC0500012766.hg.1 NR_030311.1 Refseq
Affymetrix Human Clariom D Assay PSR0500188922.hg.1 ENST00000384887.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500188922.hg.1 NR_030311.1 Refseq
Affymetrix Human Clariom D Assay PSR0500188922.hg.1 ENST00000384887.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500188922.hg.1 NR_030311.1 Refseq
Affymetrix Human Clariom D Assay PSR0500188922.hg.1 TC0500012766.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3310538 NR_030311.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3310538 NR_030311.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3310538 NR_030311.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3310538 NR_030311.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header