This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231676_s1
See other MIR573 GE Assays ›
Gene Symbol:
MIR573
Gene Name:
microRNA 573
Gene Aliases:
MIRN573, hsa-mir-573, mir-573
Chromosome Location:
Chr.4: 24520192 - 24520290 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231676_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR573
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030299.1 1 45 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA492447.1 - 1 389 Chr. 4 - 24520057 - 24520633 [-] on Build GRCh38 71
GenBankmRNA AL712496.1 - 1 75 Chr. 4 - 24519754 - 24520317 [-] on Build GRCh38 71
GenBankmRNA DA369132.1 - 1 477 Chr. 4 - 24520160 - 24520722 [-] on Build GRCh38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PPARG coactivator 1 alpha PPARGC1A LEM6,
PGC-1(alpha),
PGC-1alpha,
PGC-1v,
PGC1,
PGC1A,
PPARGC1
-47.4kb Sense Chr. 4 - 23792021 - 24472975 on NCBI Build GRCh38
Downstream DEAH-box helicase 15 DHX15 DBP1,
DDX15,
HRH2,
PRP43,
PRPF43,
PrPp43p
7.2kb Sense Chr. 4 - 24527464 - 24584603 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0400010249.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001087.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Genome CGH + SNP Microarray A_16_P16621365 - -
Agilent Human Genome CGH + SNP Microarray A_16_P16621365 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986514 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139477 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139517 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139492 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986529 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139523 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139520 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139479 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986487 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986511 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139503 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139505 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986517 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15553046 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139488 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139521 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986530 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15553050 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986491 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986486 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139514 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986538 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139484 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139497 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139493 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986519 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139483 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139508 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139471 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986507 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139512 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986502 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139506 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139486 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986524 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139472 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139465 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139506 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139486 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986487 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986524 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139479 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139503 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139483 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986514 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15553050 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139514 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139477 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986491 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139484 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986486 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986530 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139472 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139471 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986511 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139492 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986502 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139521 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139505 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139520 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986529 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139497 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139465 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139493 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139512 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139517 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986519 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139488 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139523 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986517 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15553046 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139508 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139481 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986538 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02986507 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23139470 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400010249.hg.1 ENST00000384964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010249.hg.1 NR_030299.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014955.hg.1 ENST00000384964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014955.hg.1 NR_030299.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014955.hg.1 TC04001087.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0400170129.hg.1 ENST00000384964.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400170129.hg.1 NR_030299.1 Refseq
Affymetrix Human Clariom D Assay PSR0400170129.hg.1 ENST00000384964.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400170129.hg.1 NR_030299.1 Refseq
Affymetrix Human Clariom D Assay PSR0400170129.hg.1 TC0400010249.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400010249.hg.1 ENST00000384964.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010249.hg.1 NR_030299.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3310705 NR_030299.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3310705 NR_030299.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3310705 NR_030299.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3310705 NR_030299.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0