This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231532_s1
See other MIR206 GE Assays ›
Gene Symbol:
MIR206
Gene Name:
microRNA 206
Gene Aliases:
MIRN206, miRNA206, mir-206
Chromosome Location:
Chr.6: 52144349 - 52144434 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231532_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR206
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029713.1 1 48 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream polycystic kidney and hepatic disease 1 (autosomal recessive) PKHD1 ARPKD,
FCYT,
TIGM1
-56.7kb Anti-sense Chr. 6 - 51613291 - 52087625 on NCBI Build GRCh38
Downstream microRNA 133b MIR133B MIRN133B,
miRNA133B,
mir-133b
4.5kb Sense Chr. 6 - 52148923 - 52149041 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC0600008245.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000650.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933715 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933668 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738427 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933694 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933684 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933725 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738467 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933727 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933690 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738429 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933693 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738475 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933728 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933704 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933719 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933675 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933731 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933691 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738473 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738460 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738476 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738422 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933700 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738436 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738424 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738482 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933708 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738454 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933713 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738434 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933722 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933720 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738421 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738413 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933687 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933710 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933695 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933699 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933711 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738483 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738431 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738429 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738482 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738427 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738421 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738434 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933691 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933704 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738483 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933708 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738449 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738460 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933675 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933668 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933690 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933695 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933699 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738467 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933700 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933728 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738431 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933694 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933710 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738475 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933713 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933711 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738436 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933717 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738476 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738454 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933719 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738422 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933722 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933684 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933727 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933725 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738473 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738413 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933720 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933687 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933693 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933731 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04738424 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24933715 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 8120206 - -
Affymetrix Human Transcriptome Array 2.0 TC0600008245.hg.1 ENST00000384872.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008245.hg.1 NR_029713.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007855.hg.1 ENST00000384872.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06007855.hg.1 NR_029713.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06007855.hg.1 TC06000650.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600008245.hg.1 ENST00000384872.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008245.hg.1 NR_029713.1 Refseq
Affymetrix Human Clariom D Assay PSR0600157620.hg.1 ENST00000384872.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600157620.hg.1 NR_029713.1 Refseq
Affymetrix Human Clariom D Assay PSR0600157620.hg.1 ENST00000384872.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600157620.hg.1 NR_029713.1 Refseq
Affymetrix Human Clariom D Assay PSR0600157620.hg.1 TC0600008245.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309549 NR_029713.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309549 NR_029713.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309549 NR_029713.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309549 NR_029713.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0