This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231518_s1
See other MIR129-2 GE Assays ›
Gene Symbol:
MIR129-2
Gene Name:
microRNA 129-2
Gene Aliases:
MIR-129b, MIRN129-2, mir-129-2
Chromosome Location:
Chr.11: 43581394 - 43581483 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231518_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR129-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029697.1 1 44 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MIR670 host gene MIR670HG -8.8kb Sense Chr. 11 - 43569306 - 43570400 on NCBI Build GRCh38
Downstream hydroxysteroid 17-beta dehydrogenase 12 HSD17B12 KAR,
SDR12C1
99.1kb Sense Chr. 11 - 43680593 - 43856619 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1100007376.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000366.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783291 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783258 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783260 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783292 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783274 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184412 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783253 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184349 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783263 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783299 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783241 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184404 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783268 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783286 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783235 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783210 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783244 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783226 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184407 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783209 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184380 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783224 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783267 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783252 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184356 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783289 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783246 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184389 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783228 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783295 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783297 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783212 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783302 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184359 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783250 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783222 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184397 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184410 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783256 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783208 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184338 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783240 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783266 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783233 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783223 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184371 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184399 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184419 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184340 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783273 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184341 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783216 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783272 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783237 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184347 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184406 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16439594 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184332 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184349 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783297 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184356 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783224 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783240 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783291 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184389 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184399 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783222 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783256 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184332 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184380 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783268 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783267 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184338 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783241 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184347 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783244 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783302 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184359 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783237 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783258 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783235 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783273 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184412 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184404 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184410 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783208 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783226 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783274 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783223 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783216 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783212 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783209 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783272 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184407 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184419 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184371 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184397 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783210 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783263 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783299 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783250 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783228 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783286 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783289 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783260 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783292 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184340 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783253 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184341 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783266 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783233 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783246 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783295 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16439594 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28184406 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07783252 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 7939463 - -
Affymetrix Human Transcriptome Array 2.0 TC0700009034.hg.1 ENST00000384972.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009034.hg.1 NR_029596.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11004526.hg.1 ENST00000362207.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11004526.hg.1 NR_029697.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11004526.hg.1 TC11000366.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100007376.hg.1 ENST00000362207.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007376.hg.1 NR_029697.1 Refseq
Affymetrix Human Clariom D Assay TC0700009034.hg.1 ENST00000384972.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009034.hg.1 NR_029596.1 Refseq
Affymetrix Human Clariom D Assay PSR1100152055.hg.1 ENST00000362207.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100152055.hg.1 NR_029697.1 Refseq
Affymetrix Human Clariom D Assay PSR1100152055.hg.1 ENST00000362207.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100152055.hg.1 NR_029697.1 Refseq
Affymetrix Human Clariom D Assay PSR1100152055.hg.1 TC1100007376.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100007376.hg.1 ENST00000362207.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007376.hg.1 NR_029697.1 Refseq
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3308505 NR_029697.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3308505 NR_029697.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308505 NR_029697.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308505 NR_029697.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0