This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231506_s1
See other MIR144 GE Assays ›
Gene Symbol:
MIR144
Gene Name:
microRNA 144
Gene Aliases:
MIRN144, mir-144
Chromosome Location:
Chr.17: 28861533 - 28861618 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231506_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR144
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029685.1 1 43 77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX644926.1 - 1 Chr. 17 - 28861224 - 28861734 [+] on Build GRCh38 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 451a MIR451A MIR451,
MIRN451,
hsa-mir-451,
hsa-mir-451a,
mir-451a
-93.0 Sense Chr. 17 - 28861369 - 28861440 on NCBI Build GRCh38
Downstream microRNA 4732 MIR4732 mir-4732 37.0 Sense Chr. 17 - 28861655 - 28861730 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1700010217.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001309.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838331 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16966138 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16966134 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273982 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273966 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273974 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838315 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838309 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838328 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838317 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838314 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273976 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838327 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273964 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273968 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838308 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838307 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16966137 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273981 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838332 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273978 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273966 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16966138 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16966134 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273981 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16966137 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838314 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838317 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273978 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838327 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273976 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838315 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838308 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838328 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273974 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838332 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838309 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273964 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838307 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273982 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10273968 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30838331 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 8013786 - -
Affymetrix Human Transcriptome Array 2.0 PSR17017467.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17017467.hg.1 NR_029685.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17017467.hg.1 TC17001309.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000385059.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000581873.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 NR_029685.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 NR_029970.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010217.hg.1 NR_039885.1 Refseq
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000385059.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000581873.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010217.hg.1 NR_029685.1 Refseq
Affymetrix Human Clariom D Assay TC1700010217.hg.1 NR_029970.1 Refseq
Affymetrix Human Clariom D Assay TC1700010217.hg.1 NR_039885.1 Refseq
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 NR_029685.1 Refseq
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 ENST00000384886.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 ENST00000582320.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 NR_029685.1 Refseq
Affymetrix Human Clariom D Assay PSR1700180265.hg.1 TC1700010217.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309629 NR_029685.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309629 NR_029685.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309629 NR_029685.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309629 NR_029685.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0