This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231502_s1
See other MIR140 GE Assays ›
Gene Symbol:
MIR140
Gene Name:
microRNA 140
Gene Aliases:
MIRN140, miRNA140, mir-140
Chromosome Location:
Chr.16: 69933081 - 69933180 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231502_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR140
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029681.1 1 53 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream NIN1/PSMD8 binding protein 1 homolog NOB1 ART-4,
MST158,
MSTP158,
NOB1P,
PSMD8BP1
-178.1kb Anti-sense Chr. 16 - 69741854 - 69754968 on NCBI Build GRCh38
Downstream C-type lectin domain family 18 member A CLEC18A MRCL,
MRCL1,
MRLP2
17.2kb Sense Chr. 16 - 69950233 - 69968478 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1600008245.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000573.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094931 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635013 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635006 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094934 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094908 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094894 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094917 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094895 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094918 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635025 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094899 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094911 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094909 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635024 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094896 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094893 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635005 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635030 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094923 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094932 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094890 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635022 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094888 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635026 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094921 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094920 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094936 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094926 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094938 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16908500 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094912 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094905 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16908501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094904 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635014 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094937 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094940 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094886 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094928 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094882 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635013 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094938 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094934 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094888 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094893 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094931 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094894 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094937 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094920 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094936 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094916 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16908500 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635026 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094896 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635005 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094928 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635030 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094924 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094917 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635022 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635025 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094908 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094911 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094886 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094904 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635006 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094890 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094895 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094918 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094898 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635014 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094923 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635037 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094932 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094940 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094921 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094930 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094926 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094912 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30635024 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094899 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16908501 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094882 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094909 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10094905 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 7997008 - -
Affymetrix Human Transcriptome Array 2.0 TC1600008245.hg.1 ENST00000385282.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008245.hg.1 NR_029681.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16008293.hg.1 ENST00000385282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16008293.hg.1 NR_029681.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16008293.hg.1 TC16000573.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1600008245.hg.1 ENST00000385282.2 Ensembl
Affymetrix Human Clariom D Assay TC1600008245.hg.1 NR_029681.1 Refseq
Affymetrix Human Clariom D Assay PSR1600162011.hg.1 ENST00000385282.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600162011.hg.1 NR_029681.1 Refseq
Affymetrix Human Clariom D Assay PSR1600162011.hg.1 ENST00000385282.2 Ensembl
Affymetrix Human Clariom D Assay PSR1600162011.hg.1 NR_029681.1 Refseq
Affymetrix Human Clariom D Assay PSR1600162011.hg.1 TC1600008245.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3309849 NR_029681.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3309849 NR_029681.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309849 NR_029681.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309849 NR_029681.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0