This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04231472_s1
See other MIR212 GE Assays ›
Gene Symbol:
MIR212
Gene Name:
microRNA 212
Gene Aliases:
MIRN212, mir-212
Chromosome Location:
Chr.17: 2050271 - 2050380 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04231472_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR212
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029625.1 1 54 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 132 MIR132 MIRN132,
miRNA132,
mir-132
-263.0 Sense Chr. 17 - 2049908 - 2050008 on NCBI Build GRCh38
Downstream hypermethylated in cancer 1 HIC1 ZBTB29,
ZNF901,
hic-1
4.7kb Anti-sense Chr. 17 - 2055099 - 2059687 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
This assay targets the same location as Clariom D transcript cluster(s) TC1700009415.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000987.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ArrayStar Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192462 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192469 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741423 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741426 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741421 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192465 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741423 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741417 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192469 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192470 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741421 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30741426 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192465 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192464 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10192462 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.1 ST Array 8011220 - -
Affymetrix Human Transcriptome Array 2.0 PSR17013203.hg.1 ENST00000385218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17013203.hg.1 NR_029625.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17013203.hg.1 TC17000987.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1700009415.hg.1 ENST00000586026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700009415.hg.1 NR_029625.1 Refseq
Affymetrix Human Clariom D Assay TC1700009415.hg.1 ENST00000586026.1 Ensembl
Affymetrix Human Clariom D Assay TC1700009415.hg.1 NR_029625.1 Refseq
Affymetrix Human Clariom D Assay PSR1700172079.hg.1 ENST00000586026.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700172079.hg.1 NR_029625.1 Refseq
Affymetrix Human Clariom D Assay PSR1700172079.hg.1 ENST00000586026.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700172079.hg.1 NR_029625.1 Refseq
Affymetrix Human Clariom D Assay PSR1700172079.hg.1 TC1700009415.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A054992 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A054992 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3311005 NR_029625.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3311005 NR_029625.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3311005 NR_029625.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3311005 NR_029625.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0