Assay Details
Gene Symbol: MIR204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029621.1 | 1 | 52 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transient receptor potential cation channel subfamily M member 3 | TRPM3 |
GON-2, LTRPC3, MLSN2 |
Sense | Chr. 9 - 70529063 - 71446989 on NCBI Build GRCh38 | |
Downstream | transmembrane protein 2 | TMEM2 | 873.3kb | Sense | Chr. 9 - 71683366 - 71769532 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900010376.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001203.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062567 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062582 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06724012 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06724005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062580 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062590 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062561 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16225159 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16225164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723979 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16225159 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062580 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06724005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16225164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06724012 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723966 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062582 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723976 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723974 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062567 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062561 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723979 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06723988 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062590 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27062578 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 8161695 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0900010376.hg.1 | ENST00000385200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010376.hg.1 | NR_029621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09015414.hg.1 | ENST00000385200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09015414.hg.1 | NR_029621.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09015414.hg.1 | TC09001203.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0900010376.hg.1 | ENST00000385200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010376.hg.1 | NR_029621.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171034.hg.1 | ENST00000385200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171034.hg.1 | NR_029621.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171034.hg.1 | ENST00000385200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171034.hg.1 | NR_029621.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171034.hg.1 | TC0900010376.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3308738 | NR_029621.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3308738 | NR_029621.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308738 | NR_029621.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308738 | NR_029621.1 | human_transcriptome |
