Assay Details
Gene Symbol: MIR139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029603.1 | 1 | 40 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1537 | LINC01537 | -41.8kb | Anti-sense | Chr. 11 - 72570656 - 72573222 on NCBI Build GRCh38 | |
Downstream |
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1 |
ARAP1 | CENTD2 | 69.9kb | Sense | Chr. 11 - 72685069 - 72752389 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011559.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002060.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287444 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287440 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869712 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287458 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287409 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287414 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16472392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869709 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869709 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869712 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287409 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869666 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287440 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869714 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287444 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16472392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287458 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287414 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07869703 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28287427 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 7950195 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11024640.hg.1 | ENST00000385074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11024640.hg.1 | NR_029603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11024640.hg.1 | TC11002060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011559.hg.1 | ENST00000606837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011559.hg.1 | NR_029603.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100189106.hg.1 | ENST00000606837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189106.hg.1 | NR_029603.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100189106.hg.1 | ENST00000606837.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100189106.hg.1 | NR_029603.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100189106.hg.1 | TC1100011559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100011559.hg.1 | ENST00000606837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011559.hg.1 | NR_029603.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3309644 | NR_029603.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309644 | NR_029603.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309644 | NR_029603.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309644 | NR_029603.1 | human_transcriptome |
