This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04230791_u1
See other POLB GE Assays ›
Gene Symbol:
POLB
Gene Name:
DNA polymerase beta
Gene Aliases:
-
Chromosome Location:
Chr.8: 42338452 - 42371813 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04230791_u1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002690.2 NP_002681.1 10 765 207
XM_005273535.3 XP_005273592.1 11 867 207
XM_005273536.3 XP_005273593.1 11 867 120
XM_005273537.3 XP_005273594.1 10 762 120
XM_005273538.2 XP_005273595.1 10 777 207
XM_005273539.2 XP_005273596.1 9 672 207
XM_005273540.4 XP_005273597.1 6 526 207
XM_006716353.2 XP_006716416.1 7 411 207
XM_017013583.1 XP_016869072.1 8 669 207
XM_017013584.1 XP_016869073.1 7 562 207
GenBank mRNA AK223537.1 - 10 207
AK294025.1 - 6 484 207
AK314976.1 - 10 726 207
BC100288.1 - 10 729 6151069 207
BC106909.1 - 10 691 40031807 207
CR536503.1 - 10 595 207
CR541802.1 - 10 595 207
D29013.1 - 10 708 207
L11607.1 - 10 595 207
M13140.1 - 10 708 207

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLB

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

DNA-dependent DNA replication
DNA repair
base-excision repair
base-excision repair, base-free sugar-phosphate removal
base-excision repair, gap-filling
base-excision repair, DNA ligation
pyrimidine dimer repair
nucleotide-excision repair, DNA gap filling
inflammatory response
cellular response to DNA damage stimulus
salivary gland morphogenesis
aging
intrinsic apoptotic signaling pathway in response to DNA damage
response to gamma radiation
somatic hypermutation of immunoglobulin genes
response to ethanol
lymph node development
spleen development
homeostasis of number of cells
neuron apoptotic process
response to hyperoxia
immunoglobulin heavy chain V-D-J recombination
DNA biosynthetic process
damaged DNA binding
DNA-directed DNA polymerase activity
DNA-(apurinic or apyrimidinic site) lyase activity
protein binding
microtubule binding
lyase activity
enzyme binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0