This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04189060_s1
See other RPRM GE Assays ›
Gene Symbol:
RPRM
Gene Name:
reprimo, TP53 dependent G2 arrest mediator candidate
Gene Aliases:
REPRIMO
Chromosome Location:
Chr.2: 153477338 - 153478808 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04189060_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPRM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019845.2 NP_062819.1 1 350 94
GenBank mRNA AB043585.1 - 1 350 94
AK074808.1 - 1 302 94
AK311958.1 - 1 210 94
BC002908.2 - 1 294 3942270 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014641.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002438.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPRM

Gene Ontology Categories:

Function(s) Process(es)

cell cycle arrest
regulation of mitotic cell cycle
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02038476.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02038476.hg.1 NM_019845.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02038476.hg.1 TC02002438.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 NM_019845.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 NM_019845.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 NM_019845.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200014641.hg.1 NM_019845.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02038477.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02038477.hg.1 NM_019845.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02038477.hg.1 TC02002438.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200206547.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206547.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206547.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206547.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206547.hg.1 TC0200014641.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200206550.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206550.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206550.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206550.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206550.hg.1 TC0200014641.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200206548.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206548.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206548.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206548.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206548.hg.1 TC0200014641.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200206549.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206549.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206549.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200206549.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay PSR0200206549.hg.1 TC0200014641.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014641.hg.1 ENST00000325926.3 Ensembl
Affymetrix Human Clariom D Assay TC0200014641.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay TC0200014641.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay TC0200014641.hg.1 NM_019845.2 Refseq
Affymetrix Human Clariom D Assay TC0200014641.hg.1 NM_019845.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header