This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04184583_m1
See other APOBEC3F GE Assays ›
Gene Symbol:
APOBEC3F
Gene Name:
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
Gene Aliases:
A3F, ARP8, BK150C2.4.MRNA, KA6
Chromosome Location:
Chr.22: 39020454 - 39055972 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04184583_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOBEC3F
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006666.1 NP_001006667.1 2 - 3 467 196
XM_017028643.1 XP_016884132.1 3 - 4 922 196
GenBank mRNA BM681311.1 - 1 - 2 196
CR456395.1 - 2 - 3 278 196

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
cytidine deamination
negative regulation of transposition
base conversion or substitution editing
negative regulation of viral genome replication
innate immune response
negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
negative regulation of viral process
defense response to virus
DNA cytosine deamination
DNA demethylation
RNA binding
cytidine deaminase activity
protein binding
zinc ion binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0