This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04183413_m1
See other PITX2 GE Assays ›
Gene Symbol:
PITX2
Gene Name:
paired like homeodomain 2
Gene Aliases:
ARP1, Brx1, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, PTX2, RGS, RIEG, RIEG1, RS
Chromosome Location:
Chr.4: 110617423 - 110642123 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04183413_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PITX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000325.5 NP_000316.2 1 - 2 840 97
GenBank mRNA AF048722.1 - 1 - 2 795 97
AK313987.1 - 1 - 2 495 97
BC013998.2 - 1 - 2 839 3937807 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011600.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001471.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PITX2

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

patterning of blood vessels
vasculogenesis
in utero embryonic development
neuron migration
extraocular skeletal muscle development
atrioventricular valve development
cardiac neural crest cell migration involved in outflow tract morphogenesis
pulmonary myocardium development
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
determination of left/right symmetry
myoblast fusion
male gonad development
female gonad development
response to hormone
Wnt signaling pathway
subthalamic nucleus development
hypothalamus cell migration
regulation of cell migration
embryonic camera-type eye development
response to vitamin A
embryonic hindlimb morphogenesis
hair cell differentiation
vascular smooth muscle cell differentiation
deltoid tuberosity development
odontogenesis of dentin-containing tooth
odontogenesis
camera-type eye development
positive regulation of DNA binding
positive regulation of transcription from RNA polymerase II promoter
spleen development
embryonic digestive tract morphogenesis
atrial cardiac muscle tissue morphogenesis
ventricular cardiac muscle cell development
somatotropin secreting cell differentiation
prolactin secreting cell differentiation
ventricular septum morphogenesis
left lung morphogenesis
pulmonary vein morphogenesis
superior vena cava morphogenesis
endodermal digestive tract morphogenesis
iris morphogenesis
cell proliferation involved in outflow tract morphogenesis
left/right axis specification
positive regulation of myoblast proliferation
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
chromatin DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 ENST00000306732.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 ENST00000354925.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 ENST00000355080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 ENST00000394598.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 NM_000325.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 NM_001204397.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 NM_001204398.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 NM_001204399.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 NM_153426.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 NM_153427.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04020910.hg.1 TC04001471.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000306732.7 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000354925.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000355080.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000394598.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000511837.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000511990.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000557119.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000613094.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000614423.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000616641.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_000325.5 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_001204397.1 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_001204398.1 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_001204399.1 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_153426.2 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_153427.2 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000306732.7 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000354925.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000355080.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000394598.6 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000511837.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000511990.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000557119.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000613094.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000614423.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 ENST00000616641.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_000325.5 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_001204397.1 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_001204398.1 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_001204399.1 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_153426.2 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 NM_153427.2 Refseq
Affymetrix Human Clariom D Assay PSR0400179694.hg.1 TC0400011600.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0