This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03987867_m1
See other NHSL2 GE Assays ›
Gene Symbol:
NHSL2
Gene Name:
NHS like 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 71911073 - 72153286 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03987867_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NHSL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013627.2 NP_001013649.2 6 - 7 3217 60
XM_011530934.2 XP_011529236.1 6 - 7 8009 60
XM_017029473.1 XP_016884962.1 6 - 7 6979 60
XM_017029474.1 XP_016884963.1 6 - 7 3121 60
XM_017029475.1 XP_016884964.1 2 - 3 6364 60
XM_017029476.1 XP_016884965.1 2 - 3 2481 60
XM_017029477.1 XP_016884966.1 2 - 3 7395 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00011305.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000393.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NHSL2

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X005385.hg.1 ENST00000540800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X005385.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X005385.hg.1 TC0X000393.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000373677.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000510661.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000631833.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay TC0X00011305.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00177673.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00177673.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00177673.hg.1 ENST00000632230.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00177673.hg.1 NM_001013627.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00177673.hg.1 TC0X00011305.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0