This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03924941_s1
See other LOC105374181 GE Assays ›
Gene Symbol:
LOC105374181
Gene Name:
uncharacterized LOC105374181
Gene Aliases:
-
Chromosome Location:
Chr.3: 158866998 - 158873236 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03924941_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105374181
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK130735.1 - 1 1549 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA675109.1 - 1 282 Chr. 3 - 158871162 - 158871745 [+] on Build GRCh38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream major facilitator superfamily domain containing 1 MFSD1 SMAP4 -40.2kb Sense Chr. 3 - 158801870 - 158829719 on NCBI Build GRCh38
Downstream IQ motif containing J IQCJ 197.4kb Sense Chr. 3 - 159069252 - 159266307 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009322.hg.1 ENST00000624004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009322.hg.1 ENST00000624004.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0