This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03918640_s1
See other TTLL7-IT1 GE Assays ›
Gene Symbol:
TTLL7-IT1
Gene Name:
TTLL7 intronic transcript 1
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03918640_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTLL7-IT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC042527.1 - 2 2049 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101927587 LOC101927587 -118.1kb Sense Chr. 1 - 83575787 - 83860996 on NCBI Build GRCh38
Downstream protein kinase cAMP-activated catalytic subunit beta PRKACB PKA C-beta,
PKACB
96.4kb Anti-sense Chr. 1 - 84077975 - 84238498 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100014744.hg.1 ENST00000455929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014744.hg.1 ENST00000455929.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014744.hg.1 ENST00000455929.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014744.hg.1 ENST00000455929.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0