Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196272.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196272.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196272.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196272.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196272.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196272.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196272.hg.1 |
TC1200012277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200196274.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196274.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196274.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196274.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196274.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196274.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196274.hg.1 |
TC1200012277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012277.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196270.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196270.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196270.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196270.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196270.hg.1 |
ENST00000498967.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196270.hg.1 |
NR_110049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196270.hg.1 |
TC1200012277.hg.1 |
Clariom D TC |