This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03904847_m1
See other LOC100505736 GE Assays ›
Gene Symbol:
LOC100505736
Gene Name:
uncharacterized LOC100505736
Gene Aliases:
-
Chromosome Location:
Chr.2: 28148555 - 28206961 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03904847_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100505736
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120504.1 1 - 2 474 71
GenBank mRNA AK057415.1 - 1 - 2 477 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB022519.1 - 1 - 2 474 Chr. 2 - 28151188 - 28206961 [-] on Build GRCh38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 4263 MIR4263 -154.7kb Anti-sense Chr. 2 - 27996367 - 27996449 on NCBI Build GRCh38
Downstream uncharacterized LOC100505716 LOC100505716 100.7kb Sense Chr. 2 - 28307691 - 28310459 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012104.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0200188425.hg.1 NR_120504.1 Refseq
Affymetrix Human Clariom D Assay PSR0200188425.hg.1 NR_120504.1 Refseq
Affymetrix Human Clariom D Assay PSR0200188425.hg.1 TC0200012104.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0