This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03900193_s1
See other LOC101930164 GE Assays ›
Gene Symbol:
LOC101930164
Gene Name:
uncharacterized LOC101930164
Gene Aliases:
-
Chromosome Location:
Chr.12: 40140926 - 40159940 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03900193_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101930164
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK026751.1 - 1 953 86
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE172851.1 - 1 290 Chr. 12 - 40141876 - 40142195 [-] on Build GRCh38 86

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 2 member 13 SLC2A13 HMIT -34.9kb Sense Chr. 12 - 39755021 - 40106085 on NCBI Build GRCh38
Downstream leucine rich repeat kinase 2 LRRK2 AURA17,
DARDARIN,
PARK8,
RIPK7,
ROCO2
82.1kb Anti-sense Chr. 12 - 40224895 - 40369285 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200010419.hg.1 ENST00000563933.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010419.hg.1 ENST00000563933.1 Ensembl
Affymetrix Human Clariom D Assay TC1200010419.hg.1 ENST00000563933.1 Ensembl
Affymetrix Human Clariom D Assay TC1200010419.hg.1 ENST00000563933.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0