This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03887524_s1
See other UBE2V2 GE Assays ›
Gene Symbol:
UBE2V2
Gene Name:
ubiquitin conjugating enzyme E2 V2
Gene Aliases:
DDVIT1, DDVit-1, EDAF-1, EDPF-1, EDPF1, MMS2, UEV-2, UEV2
Chromosome Location:
Chr.8: 47997527 - 48063545 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03887524_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2V2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK123928.1 - 1 1449 76
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB537509.1 - 1 55 Chr. 8 - 48064278 - 48064706 [+] on Build GRCh38 76
GenBankmRNA BX092767.1 - 1 335 Chr. 8 - 48063998 - 48064703 [+] on Build GRCh38 76
GenBankmRNA AI243174.1 - 1 Chr. 8 - 48064268 - 48064708 [-] on Build GRCh38 76
GenBankmRNA AI431959.1 - 1 Chr. 8 - 48064258 - 48064609 [-] on Build GRCh38 76
GenBankmRNA AW002051.1 - 1 Chr. 8 - 48064261 - 48064708 [-] on Build GRCh38 76
GenBankmRNA AA436186.1 - 1 Chr. 8 - 48064264 - 48064702 [-] on Build GRCh38 76
GenBankmRNA DB327904.1 - 1 Chr. 8 - 48064190 - 48064700 [-] on Build GRCh38 76
GenBankmRNA AW976892.1 - 1 Chr. 8 - 48064073 - 48064699 [-] on Build GRCh38 76
GenBankmRNA AI634267.1 - 1 Chr. 8 - 48064270 - 48064708 [-] on Build GRCh38 76
GenBankmRNA AA436311.1 - 1 336 Chr. 8 - 48063998 - 48064483 [+] on Build GRCh38 76
GenBankmRNA BU677186.1 - 1 Chr. 8 - 48064042 - 48064703 [-] on Build GRCh38 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream minichromosome maintenance complex component 4 MCM4 CDC21,
CDC54,
NKCD,
NKGCD,
P1-CDC21,
hCdc21
-84.7kb Sense Chr. 8 - 47960203 - 47978160 on NCBI Build GRCh38
Downstream uncharacterized LOC101929268 LOC101929268 486.9kb Sense Chr. 8 - 48551567 - 48698510 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0