This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03886219_s1
See other CACNG2 GE Assays ›
Gene Symbol:
CACNG2
Gene Name:
calcium voltage-gated channel auxiliary subunit gamma 2
Gene Aliases:
MRD10
Chromosome Location:
Chr.22: 36560869 - 36702645 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03886219_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CACNG2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006078.3 NP_006069.1 4 4226 69
XM_017028531.1 XP_016884020.1 3 4023 69
GenBank mRNA AK123431.1 - 1 1750 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI422240.1 - 1 Chr. 22 - 36560876 - 36561325 [+] on Build GRCh38 69
GenBankmRNA AI148157.1 - 1 Chr. 22 - 36560869 - 36561299 [+] on Build GRCh38 69
GenBankmRNA AI571218.1 - 1 Chr. 22 - 36560870 - 36561215 [+] on Build GRCh38 69
GenBankmRNA AI939486.1 - 1 Chr. 22 - 36560872 - 36561463 [+] on Build GRCh38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream eukaryotic translation initiation factor 3 subunit D EIF3D EIF3S7,
eIF3-p66,
eIF3-zeta
-31.4kb Sense Chr. 22 - 36510850 - 36529431 on NCBI Build GRCh38
Downstream uncharacterized LOC105373021 LOC105373021 141.0kb Anti-sense Chr. 22 - 36703876 - 36767089 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200008613.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CACNG2

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
neuromuscular junction development
transmission of nerve impulse
membrane depolarization
membrane hyperpolarization
cardiac conduction
calcium ion transmembrane transport
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
voltage-gated calcium channel activity
calcium channel activity
protein binding
channel regulator activity
ionotropic glutamate receptor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 ENST00000562756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay PSR2200162105.hg.1 ENST00000562756.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200162105.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay PSR2200162105.hg.1 ENST00000562756.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200162105.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay PSR2200162105.hg.1 TC2200008613.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000300105.6 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 ENST00000562756.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Affymetrix Human Clariom D Assay TC2200008613.hg.1 NM_006078.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 145100 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0