This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03871859_m1
See other LINC00536 GE Assays ›
Gene Symbol:
LINC00536
Gene Name:
long intergenic non-protein coding RNA 536
Gene Aliases:
-
Chromosome Location:
Chr.8: 115950511 - 116325059 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03871859_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00536
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046215.1 4 - 5 1334 99
GenBank mRNA BC040336.1 - 4 - 5 1334 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transcriptional repressor GATA binding 1 TRPS1 GC79,
LGCR
-460.6kb Sense Chr. 8 - 115408496 - 115669633 on NCBI Build GRCh38
Downstream eukaryotic translation initiation factor 3 subunit H EIF3H EIF3S3,
eIF3-gamma,
eIF3-p40
384.8kb Sense Chr. 8 - 116644816 - 116755823 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800011552.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay TC0800011552.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178500.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800178500.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178500.hg.1 ENST00000505156.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800178500.hg.1 NR_046215.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178500.hg.1 TC0800011552.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0